Assay Details
| Gene Symbol: tomm70a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200002.1 | NP_956296.1 | 10 - 11 | 1472 | 72 | |
| GenBank mRNA | BC059538.1 | - | 10 - 11 | 1472 | 72 | |
| BC171712.1 | - | 10 - 11 | 1572 | 72 | ||
| BC171716.1 | - | 10 - 11 | 1572 | 72 | ||
More Information
Set Membership: |
|

tomm70a
Genomic Map