Assay Details
| Gene Symbol: rpl30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200028.1 | NP_956322.1 | 244 | 90 | ||
| GenBank mRNA | BC049055.1 | - | 244 | 90 | ||
| BC062278.1 | - | 3 - 4 | 213 | 90 | ||
More Information
Set Membership: |
|

rpl30
Genomic Map