Assay Details
| Gene Symbol: vps29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200037.2 | NP_956331.1 | 3 - 4 | 531 | 137 | |
| XM_005161202.4 | XP_005161259.1 | 4 - 5 | 534 | 137 | ||
| GenBank mRNA | BC045981.1 | - | 3 - 4 | 533 | 137 | |
| BC071331.1 | - | 3 - 4 | 468 | 137 | ||
More Information
Set Membership: |
|

vps29
Genomic Map