Assay Details
| Gene Symbol: alox5ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200061.2 | NP_956355.2 | 2 - 3 | 206 | 66 | |
| GenBank mRNA | BC053181.1 | - | 2 - 3 | 66 | ||
| BC164909.1 | - | 2 - 3 | 66 | |||
More Information
Set Membership: |
|

alox5ap
Genomic Map