Assay Details
| Gene Symbol: brms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200097.1 | NP_956391.1 | 5 - 6 | 591 | 69 | |
| GenBank mRNA | BC049431.1 | - | 5 - 6 | 591 | 69 | |
| BC154789.1 | - | 5 - 6 | 635 | 69 | ||
More Information
Set Membership: |
|

brms1
Genomic Map