Assay Details
| Gene Symbol: arhgap29b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200111.2 | NP_956405.2 | 14 - 15 | 1908 | 63 | |
| XM_005163312.4 | XP_005163369.1 | 14 - 15 | 2016 | 63 | ||
| XM_005163314.4 | XP_005163371.1 | 14 - 15 | 1999 | 63 | ||
| XM_005163315.4 | XP_005163372.1 | 12 - 13 | 1829 | 63 | ||
| GenBank mRNA | BC059184.1 | - | 14 - 15 | 1901 | 63 | |
More Information
Set Membership: |
|

arhgap29b
Genomic Map