Assay Details
| Gene Symbol: arhgap29b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200111.2 | NP_956405.2 | 20 - 21 | 2863 | 100 | |
| XM_005163312.4 | XP_005163369.1 | 20 - 21 | 2971 | 100 | ||
| XM_005163314.4 | XP_005163371.1 | 20 - 21 | 2954 | 100 | ||
| XM_005163315.4 | XP_005163372.1 | 18 - 19 | 2784 | 100 | ||
| GenBank mRNA | BC059184.1 | - | 20 - 21 | 2856 | 100 | |
More Information
Set Membership: |
|

arhgap29b
Genomic Map