Assay Details
| Gene Symbol: arhgap29b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200111.2 | NP_956405.2 | 6 - 7 | 831 | 72 | |
| XM_005163312.4 | XP_005163369.1 | 6 - 7 | 939 | 72 | ||
| XM_005163315.4 | XP_005163372.1 | 4 - 5 | 752 | 72 | ||
| GenBank mRNA | BC059184.1 | - | 6 - 7 | 824 | 72 | |
More Information
Set Membership: |
|

arhgap29b
Genomic Map