Assay Details
| Gene Symbol: arhgap29b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200111.2 | NP_956405.2 | 7 - 8 | 981 | 100 | |
| XM_005163312.4 | XP_005163369.1 | 7 - 8 | 1089 | 100 | ||
| XM_005163314.4 | XP_005163371.1 | 7 - 8 | 1072 | 100 | ||
| XM_005163315.4 | XP_005163372.1 | 5 - 6 | 902 | 100 | ||
| GenBank mRNA | BC059184.1 | - | 7 - 8 | 974 | 100 | |
More Information
Set Membership: |
|

arhgap29b
Genomic Map