Assay Details
| Gene Symbol: arhgap29b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200111.2 | NP_956405.2 | 8 - 9 | 1050 | 66 | |
| XM_005163312.4 | XP_005163369.1 | 8 - 9 | 1158 | 66 | ||
| XM_005163314.4 | XP_005163371.1 | 8 - 9 | 1141 | 66 | ||
| XM_005163315.4 | XP_005163372.1 | 6 - 7 | 971 | 66 | ||
| GenBank mRNA | BC059184.1 | - | 8 - 9 | 1043 | 66 | |
More Information
Set Membership: |
|

arhgap29b
Genomic Map