Assay Details
| Gene Symbol: rundc3ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200149.1 | NP_956443.1 | 11 - 12 | 1349 | 74 | |
| GenBank mRNA | BC044502.1 | - | 11 - 12 | 1349 | 74 | |
| BC044540.1 | - | 11 - 12 | 1349 | 74 | ||
| BC155785.1 | - | 10 - 11 | 1532 | 74 | ||
| BC165804.1 | - | 10 - 11 | 1178 | 74 | ||
| EF646262.1 | - | 10 - 11 | 1158 | 74 | ||
More Information
Set Membership: |
|

rundc3ab
Genomic Map