Assay Details
| Gene Symbol: rundc3ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200149.1 | NP_956443.1 | 2 - 3 | 312 | 76 | |
| GenBank mRNA | BC044502.1 | - | 2 - 3 | 312 | 76 | |
| BC044540.1 | - | 2 - 3 | 312 | 76 | ||
| BC155785.1 | - | 1 - 2 | 495 | 76 | ||
| BC165804.1 | - | 1 - 2 | 141 | 76 | ||
| EF646262.1 | - | 1 - 2 | 121 | 76 | ||
More Information
Set Membership: |
|

rundc3ab
Genomic Map