Assay Details
| Gene Symbol: rundc3ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200149.1 | NP_956443.1 | 3 - 4 | 433 | 142 | |
| GenBank mRNA | BC044502.1 | - | 3 - 4 | 433 | 142 | |
| BC044540.1 | - | 3 - 4 | 433 | 142 | ||
| BC165804.1 | - | 2 - 3 | 262 | 142 | ||
More Information
Set Membership: |
|

rundc3ab
Genomic Map