Assay Details
| Gene Symbol: rundc3ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200149.1 | NP_956443.1 | 9 - 10 | 1133 | 102 | |
| GenBank mRNA | BC044502.1 | - | 9 - 10 | 1133 | 102 | |
| BC044540.1 | - | 9 - 10 | 1133 | 102 | ||
| BC155785.1 | - | 8 - 9 | 102 | |||
| BC165804.1 | - | 8 - 9 | 962 | 102 | ||
| EF646262.1 | - | 8 - 9 | 942 | 102 | ||
More Information
Set Membership: |
|

rundc3ab
Genomic Map