Assay Details
| Gene Symbol: kbtbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200209.1 | NP_956503.1 | 2 - 3 | 562 | 69 | |
| GenBank mRNA | BC045989.1 | - | 2 - 3 | 562 | 69 | |
| BC152675.1 | - | 1 - 2 | 937 | 69 | ||
More Information
Set Membership: |
|

kbtbd4
Genomic Map