Assay Details
| Gene Symbol: gde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200243.1 | NP_956537.1 | 5 - 6 | 1046 | 73 | |
| GenBank mRNA | BC047818.1 | - | 5 - 6 | 1046 | 73 | |
| BC065614.1 | - | 5 - 6 | 966 | 73 | ||
| BC154208.1 | - | 5 - 6 | 1064 | 73 | ||
More Information
Set Membership: |
|

gde1
Genomic Map