Assay Details
| Gene Symbol: ptgesl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200280.1 | NP_956574.1 | 3 - 4 | 597 | 125 | |
| XM_021478331.1 | XP_021334006.1 | 3 - 4 | 268 | 125 | ||
| XM_021478332.1 | XP_021334007.1 | 3 - 4 | 591 | 125 | ||
| GenBank mRNA | BC049325.1 | - | 3 - 4 | 597 | 125 | |
| BC171675.1 | - | 3 - 4 | 580 | 125 | ||
More Information
Set Membership: |
|

ptgesl
Genomic Map