Assay Details
| Gene Symbol: ptgesl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200280.1 | NP_956574.1 | 4 - 5 | 745 | 70 | |
| XM_021478331.1 | XP_021334006.1 | 4 - 5 | 416 | 70 | ||
| GenBank mRNA | BC049325.1 | - | 4 - 5 | 745 | 70 | |
| BC171675.1 | - | 4 - 5 | 728 | 70 | ||
More Information
Set Membership: |
|

ptgesl
Genomic Map