Assay Details
| Gene Symbol: ptgesl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200280.1 | NP_956574.1 | 6 - 7 | 1072 | 76 | |
| XM_021478331.1 | XP_021334006.1 | 6 - 7 | 743 | 76 | ||
| GenBank mRNA | BC049325.1 | - | 6 - 7 | 1072 | 76 | |
| BC150469.1 | - | 5 - 6 | 1153 | 76 | ||
| BC171675.1 | - | 6 - 7 | 1055 | 76 | ||
More Information
Set Membership: |
|

ptgesl
Genomic Map