Assay Details
| Gene Symbol: kyat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200344.2 | NP_956638.2 | 5 - 6 | 641 | 145 | |
| GenBank mRNA | BC053152.1 | - | 5 - 6 | 604 | 145 | |
More Information
Set Membership: |
|

kyat3
Genomic Map