Assay Details
| Gene Symbol: kyat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200344.2 | NP_956638.2 | 7 - 8 | 891 | 101 | |
| XM_005168757.4 | XP_005168814.1 | 7 - 8 | 721 | 101 | ||
| XM_005168758.4 | XP_005168815.1 | 7 - 8 | 802 | 101 | ||
| XM_021476800.1 | XP_021332475.1 | 5 - 6 | 559 | 101 | ||
| XM_021476801.1 | XP_021332476.1 | 6 - 7 | 588 | 101 | ||
| GenBank mRNA | BC053152.1 | - | 7 - 8 | 854 | 101 | |
More Information
Set Membership: |
|

kyat3
Genomic Map