Assay Details
| Gene Symbol: zgc:63972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200354.1 | NP_956648.1 | 2 - 3 | 332 | 61 | |
| GenBank mRNA | BC053175.1 | - | 2 - 3 | 332 | 61 | |
| BC155752.1 | - | 2 - 3 | 324 | 61 | ||
More Information
Set Membership: |
|

zgc:63972
Genomic Map