Assay Details
| Gene Symbol: sri | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200373.1 | NP_956667.1 | 7 - 8 | 638 | 69 | |
| XM_005157861.3 | XP_005157918.1 | 7 - 8 | 915 | 69 | ||
| GenBank mRNA | BC053237.1 | - | 7 - 8 | 638 | 69 | |
More Information
Set Membership: |
|

sri
Genomic Map