Assay Details
| Gene Symbol: dnajc5gb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200400.1 | NP_956694.1 | 6 - 7 | 768 | 92 | |
| GenBank mRNA | BC054133.1 | - | 6 - 7 | 768 | 92 | |
More Information
Set Membership: |
|

dnajc5gb
Genomic Map