Assay Details
| Gene Symbol: znf800b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200436.1 | NP_956730.1 | 5 - 6 | 1631 | 67 | |
| GenBank mRNA | BC054916.1 | - | 5 - 6 | 1631 | 67 | |
More Information
Set Membership: |
|

znf800b
Genomic Map