Assay Details
| Gene Symbol: capn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200445.1 | NP_956739.1 | 14 - 15 | 1690 | 95 | |
| XM_009307548.3 | XP_009305823.1 | 14 - 15 | 1767 | 95 | ||
| GenBank mRNA | BC054941.1 | - | 14 - 15 | 1690 | 95 | |
More Information
Set Membership: |
|

capn1a
Genomic Map