Assay Details
| Gene Symbol: capn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200445.1 | NP_956739.1 | 20 - 21 | 2141 | 68 | |
| XM_009307548.3 | XP_009305823.1 | 20 - 21 | 2218 | 68 | ||
| GenBank mRNA | BC054941.1 | - | 20 - 21 | 2141 | 68 | |
More Information
Set Membership: |
|

capn1a
Genomic Map