Assay Details
| Gene Symbol: camkvb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200450.1 | NP_956744.1 | 10 - 11 | 1130 | 85 | |
| GenBank mRNA | BC055127.1 | - | 10 - 11 | 1130 | 85 | |
More Information
Set Membership: |
|

camkvb
Genomic Map