Assay Details
| Gene Symbol: camkvb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200450.1 | NP_956744.1 | 4 - 5 | 494 | 75 | |
| GenBank mRNA | AY450361.1 | - | 3 - 4 | 304 | 75 | |
| BC055127.1 | - | 4 - 5 | 494 | 75 | ||
More Information
Set Membership: |
|

camkvb
Genomic Map