Assay Details
| Gene Symbol: camkvb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200450.1 | NP_956744.1 | 7 - 8 | 831 | 149 | |
| GenBank mRNA | AY450361.1 | - | 6 - 7 | 641 | 149 | |
| BC055127.1 | - | 7 - 8 | 831 | 149 | ||
More Information
Set Membership: |
|

camkvb
Genomic Map