Assay Details
| Gene Symbol: tiprl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328426.1 | NP_001315355.1 | 4 - 5 | 655 | 71 | |
| GenBank mRNA | BC057515.1 | - | 4 - 5 | 71 | ||
| BC165159.1 | - | 3 - 4 | 71 | |||
More Information
Set Membership: |
|

tiprl
Genomic Map