Assay Details
| Gene Symbol: tiprl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328426.1 | NP_001315355.1 | 5 - 6 | 784 | 81 | |
| GenBank mRNA | BC057515.1 | - | 5 - 6 | 568 | 81 | |
| BC165159.1 | - | 4 - 5 | 560 | 81 | ||
More Information
Set Membership: |
|

tiprl
Genomic Map