Assay Details
| Gene Symbol: swap70a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001362396.1 | NP_001349325.1 | 3 - 4 | 498 | 96 | |
| GenBank mRNA | BC058058.1 | - | 498 | 96 | ||
| HM752768.1 | - | 2 - 3 | 324 | 96 | ||
More Information
Set Membership: |
|

swap70a
Genomic Map