Assay Details
| Gene Symbol: grap2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200678.2 | NP_956972.1 | 4 - 5 | 453 | 103 | |
| GenBank mRNA | BC058307.1 | - | 359 | 103 | ||
| BC066707.1 | - | 4 - 5 | 333 | 103 | ||
| EH590682.1 | - | 4 - 5 | 474 | 103 | ||
More Information
Set Membership: |
|

grap2b
Genomic Map