Assay Details
| Gene Symbol: btf3l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200694.1 | NP_956988.1 | 4 - 5 | 446 | 103 | |
| XM_009305886.2 | XP_009304161.1 | 5 - 6 | 536 | 103 | ||
| XM_009305887.3 | XP_009304162.1 | 4 - 5 | 463 | 103 | ||
| XM_021479564.1 | XP_021335239.1 | 5 - 6 | 536 | 103 | ||
| GenBank mRNA | BC059432.1 | - | 4 - 5 | 446 | 103 | |
More Information
Set Membership: |
|

btf3l4
Genomic Map