Assay Details
| Gene Symbol: sap18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200720.2 | NP_957014.1 | 2 - 3 | 297 | 68 | |
| GenBank mRNA | BC059482.1 | - | 2 - 3 | 235 | 68 | |
| BC090729.1 | - | 1 - 2 | 1115 | 68 | ||
| BC155197.1 | - | 2 - 3 | 245 | 68 | ||
| EH440312.1 | - | 2 - 3 | 232 | 68 | ||
More Information
Set Membership: |
|

sap18
Genomic Map