Assay Details
| Gene Symbol: rdh8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200788.2 | NP_957082.2 | 5 - 6 | 802 | 62 | |
| GenBank mRNA | AY306006.1 | - | 5 - 6 | 800 | 62 | |
| BC059640.1 | - | 5 - 6 | 802 | 62 | ||
More Information
Set Membership: |
|

rdh8b
Genomic Map