Assay Details
| Gene Symbol: arr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200792.1 | NP_957086.1 | 7 - 8 | 460 | 127 | |
| GenBank mRNA | BC059650.1 | - | 7 - 8 | 460 | 127 | |
More Information
Set Membership: |
|

arr3b
Genomic Map