Assay Details
| Gene Symbol: ppp1r14ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200810.1 | NP_957104.1 | 1 - 2 | 204 | 127 | |
| GenBank mRNA | BC059686.1 | - | 1 - 2 | 204 | 127 | |
More Information
Set Membership: |
|

ppp1r14ab
Genomic Map