Assay Details
| Gene Symbol: ca7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200813.1 | NP_957107.1 | 2 - 3 | 327 | 124 | |
| GenBank mRNA | BC059693.1 | - | 2 - 3 | 327 | 124 | |
| BC154317.1 | - | 2 - 3 | 431 | 124 | ||
| BC171412.1 | - | 2 - 3 | 286 | 124 | ||
| BC171416.1 | - | 2 - 3 | 286 | 124 | ||
More Information
Set Membership: |
|

ca7
Genomic Map