Assay Details
| Gene Symbol: rasa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099451.1 | NP_001092921.1 | 15 - 16 | 1653 | 59 | |
| XM_005171995.3 | XP_005172052.1 | 15 - 16 | 1903 | 59 | ||
More Information
Set Membership: |
|

rasa4
Genomic Map