Assay Details
| Gene Symbol: cacna1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114548.1 | NP_001108020.1 | 18 - 19 | 2267 | 57 | |
| XM_009301497.3 | XP_009299772.1 | 19 - 20 | 2948 | 57 | ||
| XM_009301500.3 | XP_009299775.1 | 19 - 20 | 2312 | 57 | ||
| XM_009301504.3 | XP_009299779.1 | 19 - 20 | 2312 | 57 | ||
| XM_009301505.3 | XP_009299780.1 | 18 - 19 | 2906 | 57 | ||
| XM_017356096.2 | XP_017211585.1 | 18 - 19 | 2270 | 57 | ||
| XM_021476173.1 | XP_021331848.1 | 19 - 20 | 2312 | 57 | ||
| XM_021476174.1 | XP_021331849.1 | 19 - 20 | 2312 | 57 | ||
| XM_021476175.1 | XP_021331850.1 | 19 - 20 | 2312 | 57 | ||
| XM_021476176.1 | XP_021331851.1 | 19 - 20 | 2312 | 57 | ||
| XM_021476178.1 | XP_021331853.1 | 19 - 20 | 2312 | 57 | ||
| XM_021476179.1 | XP_021331854.1 | 19 - 20 | 2312 | 57 | ||
| XM_021476180.1 | XP_021331855.1 | 19 - 20 | 2312 | 57 | ||
| GenBank mRNA | KC192784.1 | - | 19 - 20 | 2312 | 57 | |
More Information
Set Membership: |
|

cacna1ba
Genomic Map