Assay Details
| Gene Symbol: cacna1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114548.1 | NP_001108020.1 | 32 - 33 | 4583 | 87 | |
| XM_009301505.3 | XP_009299780.1 | 32 - 33 | 5336 | 87 | ||
| XM_017356097.2 | XP_017211586.1 | 34 - 35 | 5348 | 87 | ||
| XM_021476174.1 | XP_021331849.1 | 35 - 36 | 4775 | 87 | ||
| XM_021476178.1 | XP_021331853.1 | 33 - 34 | 4742 | 87 | ||
| XM_021476179.1 | XP_021331854.1 | 33 - 34 | 4742 | 87 | ||
| XM_021476182.1 | XP_021331857.1 | 32 - 33 | 5315 | 87 | ||
More Information
Set Membership: |
|

cacna1ba
Genomic Map