Assay Details
| Gene Symbol: cacna1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114548.1 | NP_001108020.1 | 36 - 37 | 5012 | 90 | |
| XM_009301497.3 | XP_009299772.1 | 40 - 41 | 5846 | 90 | ||
| XM_009301500.3 | XP_009299775.1 | 38 - 39 | 5177 | 90 | ||
| XM_009301501.3 | XP_009299776.1 | 39 - 40 | 5147 | 90 | ||
| XM_009301504.3 | XP_009299779.1 | 38 - 39 | 5177 | 90 | ||
| XM_009301505.3 | XP_009299780.1 | 36 - 37 | 5765 | 90 | ||
| XM_017356095.2 | XP_017211584.1 | 40 - 41 | 5183 | 90 | ||
| XM_017356096.2 | XP_017211585.1 | 39 - 40 | 5168 | 90 | ||
| XM_017356097.2 | XP_017211586.1 | 38 - 39 | 5777 | 90 | ||
| XM_021476173.1 | XP_021331848.1 | 40 - 41 | 5210 | 90 | ||
| XM_021476174.1 | XP_021331849.1 | 39 - 40 | 5204 | 90 | ||
| XM_021476175.1 | XP_021331850.1 | 39 - 40 | 5198 | 90 | ||
| XM_021476176.1 | XP_021331851.1 | 39 - 40 | 5189 | 90 | ||
| XM_021476178.1 | XP_021331853.1 | 37 - 38 | 5171 | 90 | ||
| XM_021476179.1 | XP_021331854.1 | 37 - 38 | 5171 | 90 | ||
| XM_021476180.1 | XP_021331855.1 | 40 - 41 | 5210 | 90 | ||
| XM_021476181.1 | XP_021331856.1 | 37 - 38 | 5750 | 90 | ||
| XM_021476182.1 | XP_021331857.1 | 36 - 37 | 5744 | 90 | ||
| GenBank mRNA | KC192784.1 | - | 38 - 39 | 5177 | 90 | |
More Information
Set Membership: |
|

cacna1ba
Genomic Map