Assay Details
| Gene Symbol: cacna1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114548.1 | NP_001108020.1 | 3 - 4 | 508 | 72 | |
| XM_009301497.3 | XP_009299772.1 | 3 - 4 | 1147 | 72 | ||
| XM_009301500.3 | XP_009299775.1 | 3 - 4 | 511 | 72 | ||
| XM_009301501.3 | XP_009299776.1 | 3 - 4 | 511 | 72 | ||
| XM_009301504.3 | XP_009299779.1 | 3 - 4 | 511 | 72 | ||
| XM_009301505.3 | XP_009299780.1 | 3 - 4 | 1147 | 72 | ||
| XM_017356095.2 | XP_017211584.1 | 3 - 4 | 511 | 72 | ||
| XM_017356096.2 | XP_017211585.1 | 3 - 4 | 511 | 72 | ||
| XM_017356097.2 | XP_017211586.1 | 3 - 4 | 1147 | 72 | ||
| XM_021476173.1 | XP_021331848.1 | 3 - 4 | 511 | 72 | ||
| XM_021476174.1 | XP_021331849.1 | 3 - 4 | 511 | 72 | ||
| XM_021476175.1 | XP_021331850.1 | 3 - 4 | 511 | 72 | ||
| XM_021476176.1 | XP_021331851.1 | 3 - 4 | 511 | 72 | ||
| XM_021476178.1 | XP_021331853.1 | 3 - 4 | 511 | 72 | ||
| XM_021476179.1 | XP_021331854.1 | 3 - 4 | 511 | 72 | ||
| XM_021476180.1 | XP_021331855.1 | 3 - 4 | 511 | 72 | ||
| XM_021476181.1 | XP_021331856.1 | 3 - 4 | 1147 | 72 | ||
| XM_021476182.1 | XP_021331857.1 | 3 - 4 | 1147 | 72 | ||
| GenBank mRNA | KC192784.1 | - | 3 - 4 | 511 | 72 | |
More Information
Set Membership: |
|

cacna1ba
Genomic Map