Assay Details
| Gene Symbol: igf2bp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114558.2 | NP_001108030.2 | 10 - 11 | 1221 | 127 | |
| XM_005167469.4 | XP_005167526.1 | 10 - 11 | 1202 | 127 | ||
| XM_009304504.3 | XP_009302779.1 | 10 - 11 | 1971 | 127 | ||
| GenBank mRNA | BC155087.1 | - | 2 - 3 | 206 | 127 | |
| DQ998838.1 | - | 10 - 11 | 1221 | 127 | ||
More Information
Set Membership: |
|

igf2bp2a
Genomic Map