Assay Details
| Gene Symbol: igf2bp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114558.2 | NP_001108030.2 | 12 - 13 | 1396 | 99 | |
| XM_005167469.4 | XP_005167526.1 | 12 - 13 | 1377 | 99 | ||
| XM_009304504.3 | XP_009302779.1 | 12 - 13 | 2143 | 99 | ||
| GenBank mRNA | BC155087.1 | - | 4 - 5 | 378 | 99 | |
| DQ998838.1 | - | 12 - 13 | 1396 | 99 | ||
| EU296401.2 | - | 12 - 13 | 1603 | 99 | ||
| FJ418164.1 | - | 12 - 13 | 1603 | 99 | ||
| FJ418165.1 | - | 11 - 12 | 1299 | 99 | ||
| FJ418166.1 | - | 11 - 12 | 1299 | 99 | ||
More Information
Set Membership: |
|

igf2bp2a
Genomic Map