Assay Details
| Gene Symbol: kif2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114593.1 | NP_001108065.1 | 15 - 16 | 1668 | 66 | |
| XM_005163472.4 | XP_005163529.1 | 16 - 17 | 1740 | 66 | ||
| GenBank mRNA | BC155585.1 | - | 15 - 16 | 1668 | 66 | |
More Information
Set Membership: |
|

kif2c
Genomic Map