Assay Details
| Gene Symbol: dnase2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114738.1 | NP_001108210.1 | 4 - 5 | 432 | 66 | |
| XM_021466569.1 | XP_021322244.1 | 4 - 5 | 482 | 66 | ||
| GenBank mRNA | BC159268.1 | - | 4 - 5 | 442 | 66 | |
More Information
Set Membership: |
|

dnase2
Genomic Map