Assay Details
| Gene Symbol: sptlc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114741.1 | NP_001108213.1 | 11 - 12 | 1594 | 60 | |
| GenBank mRNA | BC159260.1 | - | 11 - 12 | 1594 | 60 | |
More Information
Set Membership: |
|

sptlc2b
Genomic Map